Order Entry
Belgium
Orders LinkContactUsLinkComponent
Electrodes, 2-Needle Array™

Electrodes, 2-Needle Array™

2-Needle Array™ handle, 5 mm gap
Catalog # 732-0030
Supplier:   BTX
Electrodes, 2-Needle Array™
Catalog # 732-0030
Supplier:  BTX
Electrodes, 2-Needle Array™
Electrodes, 2-Needle Array™

2-Needle Array™ handle, 5 mm gap

Price
360,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

2-Needle Array™ electrodes are needle-style electrodes specifically designed for in vivo drug or gene delivery applications. The electrode consists of a reusable 2-Needle Array™ handle and sterile, disposable, 2-needle arrays.

  • Stainless steel electrode, delrin handle
  • Simple, inexpensive and safe non viral technique for in vivo gene transfer by the direct injection of plasmid DNA into muscle

The electrode is available in two sizes. The 5 mm 2-needle array and handle is recommended for small muscle masses such as mouse tibialis. The 10 mm 2-needle array and handle is recommended for larger muscle masses such as rat gastronemus.

Generator compatibility: ECM® 830, ECM® 2001
Voltage range: 0 - 500 V

Pulse length range: 10 µsec to 99 msec in PBS
Handle length: 8 cm
Needle length: 20 mm

Pulse length range: 10 µsec to 99 msec in PBS

Handle length: 8 cm

Specifications
  • Description:
    2-Needle Array™ handle, 5 mm gap
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.