Order Entry
Belgium
Orders LinkContactUsLinkComponent
Electrodes, 2-Needle Array™

Electrodes, 2-Needle Array™

2-Needle Array™ handle, 10 mm gap
Catalog # 732-0029
Supplier:   BTX
Electrodes, 2-Needle Array™
Catalog # 732-0029
Supplier:  BTX
Electrodes, 2-Needle Array™
Electrodes, 2-Needle Array™

2-Needle Array™ handle, 10 mm gap

Price
394,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

2-Needle Array™ electrodes are needle-style electrodes specifically designed for in vivo drug or gene delivery applications. The electrode consists of a reusable 2-Needle Array™ handle and sterile, disposable, 2-needle arrays.

  • Stainless steel electrode, delrin handle
  • Simple, inexpensive and safe non viral technique for in vivo gene transfer by the direct injection of plasmid DNA into muscle

The electrode is available in two sizes. The 5 mm 2-needle array and handle is recommended for small muscle masses such as mouse tibialis. The 10 mm 2-needle array and handle is recommended for larger muscle masses such as rat gastronemus.

Generator compatibility: ECM® 830, ECM® 2001
Voltage range: 0 - 500 V

Pulse length range: 10 µsec to 99 msec in PBS
Handle length: 8 cm
Needle length: 20 mm

Pulse length range: 10 µsec to 99 msec in PBS

Handle length: 8 cm

Specifications
  • Description:
    2-Needle Array™ handle, 10 mm gap
  • Seq:
    01