Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Sample dippers

Sampling dipper with reinforced rim and spout, PTFE/steel, 125 mm, 80 mm, 500 ml
  BRND90454
 :   Brand
Sample dippers
  BRND90454
 :  Brand
Sample dippers
Sample dippers

Sampling dipper with reinforced rim and spout, PTFE/steel, 125 mm, 80 mm, 500 ml

 
237,00 €
   
1 * 1 ST
 

Sampling dipper with reinforced rim and spout.

  • PTFE handle with steel core

Optional extension rod for increased handle length.

 
  • Ø:
    80 mm
  • Capacity:
    500 ml
  • Description:
    Sampling dipper with reinforced rim and spout
  • Length:
    125 mm
  • Material:
    PTFE/steel
Accessories

 

  •  
    Description
     
     
  • BRND90470
    Extension rod for sampling device, 600 mm
       
    1 * 1 ST - 121,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.