Seals, PTFE, for Dispensette® and Digital Burette III 1 * 5 items
Catalog # BRND6696
Supplier: Brand
Seals, PTFE, for Dispensette® and Digital Burette III 1 * 5 items
Catalog # BRND6696
Supplier: Brand
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Schutzfilter 5 ml für Eppendorf Research Plus 1 * 50 ST
Supplier: EPPENDORF
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Accessories for Dispensette®
Supplier: Brand
Accessories for accu-jet® pro
Supplier: Brand
JUG 1000ML 31,5MM GREEN GLAS 1 * 1.183 items
Supplier: PONT PACKAGING
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
VWR®, Accessories for Mechanical Pipettes
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Measuring Cylinders, Tall Form, Class A, Borosilicate Glass
Supplier: VWR Collection
Accessories for Research® Plus pipettes
Supplier: EPPENDORF
Accessories for electronic dispensers, Multipette E3/E3x
Supplier: EPPENDORF
Vacuum seal PTFE / FKM
Supplier: Heidolph Instruments GmbH & Co.KG
Accessories for PIPETBOY acu / pro
Supplier: Integra