Orders LinkContactUsLinkComponent
Staining trough
Staining trough
Staining trough, transparent, 83×101×70 mm
Catalog # BRND474300
Supplier:   Brand
Staining trough
Catalog # BRND474300
Supplier:  Brand
Staining trough
Staining trough
Staining trough, transparent, 83×101×70 mm
Price
173,00 €
Log in for availability
1 * 4 ST
Product Details & Documents
PMP, transparent.
Trough comes with two lids: one to help protect against evaporation when not in use; one with a handle slot facilitating the staining process.
PP tray with fold down handle can accomodate 20 slides. The close fitting lid helps protect against evaporation.
Ordering information: Trough and tray (insert) have to be ordered separately.
Specifications
  • Description:
    Staining trough, transparent
  • W×D×H:
    83×101×70 mm