Orders LinkContactUsLinkComponent
Laboratory trays
300×240×70 mm
Catalog # BRND156642
Supplier:   Brand
Laboratory trays
Catalog # BRND156642
Supplier:  Brand
Laboratory trays
Laboratory trays
300×240×70 mm
Price
21,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
PP, white.
  • Very good chemical resistance
  • Very good thermal resistance
  • Smooth surfaces for easy cleaning
With ribs on base. Highly versatile trays, ideal for a wide range of applications. The ribs on the base make these trays ideal for use as developing trays.
Specifications
  • W×D×H:
    300×240×70 mm