Laboratory trays
225×180×45 mm
Catalog # BRND156640
Supplier: Brand
Jump to:
Product Details & Documents
PP, white.
- Very good chemical resistance
- Very good thermal resistance
- Smooth surfaces for easy cleaning
With ribs on base. Highly versatile trays, ideal for a wide range of applications. The ribs on the base make these trays ideal for use as developing trays.
Specifications
- W×D×H:225×180×45 mm