Order Entry
Belgium
Orders LinkContactUsLinkComponent
Dextrin

Dextrin

Plastic bottle, 500 g
Catalog # ACRO406285000
Dextrin
Catalog # ACRO406285000
CAS Number:  9004-53-9
Dextrin
Dextrin

Plastic bottle, 500 g

CAS Number:  9004-53-9
Price
95,90 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Dextrin
  • CAS Number:9004-53-9
  • Formula:(C₆H₁₂O₆)n
  • MDL Number:MFCD00081554
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,02971
  • Density:1,45 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:232-675-4

Documents

Specifications

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Sulfated ash:
    ≤0.5 %
  • Titration with HClO4:
    ≥96.0 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g

Specifications