Order Entry
Belgium
Orders LinkContactUsLinkComponent
Dextrin

Dextrin

Plastic bottle, 100 g
Catalog # ACRO406281000
Dextrin
Catalog # ACRO406281000
CAS Number:  9004-53-9
Dextrin
Dextrin

Plastic bottle, 100 g

CAS Number:  9004-53-9
Price
32,80 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Dextrin
  • CAS Number:9004-53-9
  • Formula:(C₆H₁₂O₆)n
  • MDL Number:MFCD00081554
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,02971
  • Density:1,45 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:232-675-4

Documents

Specifications

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Sulfated ash:
    ≤0.5 %
  • Titration with HClO4:
    ≥96.0 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.