Order Entry
Belgium
Orders LinkContactUsLinkComponent
Stirring rods

Stirring rods

Stirring rod, with spatula ends, PTFE, 8 mm, 250 mm
Catalog # BRND136220
Supplier:   Brand
Stirring rods
Catalog # BRND136220
Supplier:  Brand
Stirring rods
Stirring rods

Stirring rod, with spatula ends, PTFE, 8 mm, 250 mm

Price
19,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Ø:
    8 mm
  • Description:
    Stirring rod, with spatula ends
  • Length:
    250 mm
  • Material:
    PTFE
  • Seq:
    002