Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Dropping bottles, PTFE

50 ml, 100 mm, 43 mm
Catalog # BRND125328
Supplier:   Brand
Dropping bottles, PTFE
Catalog # BRND125328
Supplier:  Brand
Dropping bottles, PTFE
Dropping bottles, PTFE

50 ml, 100 mm, 43 mm

Price
125,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

PTFE, white.

  • Flexible wall for easy use
  • Screw fit cap
  • Guaranteed chemically resistant and inert

Suitable for dropping out aggressive chemicals.

Specifications
  • Ext.Ø:
    43 mm
  • Capacity:
    50 ml
  • Height:
    100 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.