Order Entry
Belgium
Orders LinkContactUsLinkComponent
Dropping bottles, PTFE

Dropping bottles, PTFE

25 ml, 93 mm, 33 mm
Catalog # BRND125316
Supplier:   Brand
Dropping bottles, PTFE
Catalog # BRND125316
Supplier:  Brand
Dropping bottles, PTFE
Dropping bottles, PTFE

25 ml, 93 mm, 33 mm

Price
113,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

PTFE, white.

  • Flexible wall for easy use
  • Screw fit cap
  • Guaranteed chemically resistant and inert

Suitable for dropping out aggressive chemicals.

Specifications
  • Ext.Ø:
    33 mm
  • Capacity:
    25 ml
  • Height:
    93 mm