Cryogenic tubes
4,00 ml, 12,5×76 mm, Yes
Catalog # 479-1205
Supplier: Brand
Jump to:
Product Details & Documents
PP, graduated. Designed for storage of biological material, such as microorganisms, human and animal cells, more. in the gaseous phase of liquid nitrogen.
- Large frosted marking area and coloured cap inserts for easy sample identification
- Temperature stability to −196 °C and autoclavable at 121 °C (2 bar), acc. DIN EN 285
- γ-ray sterile (SAL 10⁻⁶)
- Cryogenic tubes are sterile, RNase-, DNase-, DNA- and endotoxin-free
- Tubes without ring stands can be centrifuged at up to an RCF of 14000×g
Freestanding
* with round bottom
Documents
Specifications
- Cap included:Yes
- Ø×H:12,5×76 mm
- Capacity:4,00 ml
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



