3,3',5,5'-Tetramethylbenzidine (TMB) QUICK, liquid substrate for ELISA
Glass bottle, 50 mL
Supplier: Thermo Fisher Scientific
3,3',5,5'-Tetramethylbenzidine (TMB) QUICK, liquid substrate for ELISA
Catalog # ACRO328050500
Supplier: Thermo Fisher Scientific
CAS Number: 54827-17-7
Jump to:
Product Details & Documents
Synonyms: TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
- CAS Number:54827-17-7
- Formula:C₁₆H₂₀N₂
- MDL Number:MFCD00007748
- Molecular Weight:240.35 g/mol
- Storage Temperature:Refrigerator
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow-green to light blue Solution
- pH:3.50 to 3.90
- Activity:90 to 110 %
- Absorbance:≤0.025 (450/620 nm, 0.2M H2SO4)
Specifications
- Pack type:Glass bottle
- Pk:50 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



