Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Bellow connectors

29/32 NS, 120 mm, 29/32 NS
  BOHLH906-12
 :   Bohlender
Bellow connectors
  BOHLH906-12
 :  Bohlender
Bellow connectors
Bellow connectors

29/32 NS, 120 mm, 29/32 NS

 
82,90 €
   
1 * 1 ST
 

Bellow connectors, made of PTFE, with sharp folds and sealing rings on the outside of the cone.

  • Universal chemical resistance
  • Suitable for vacuum applications
  • Temperature resistance from −200 to +250 °C
  • Compensates vibrations from vacuum pumps
  • Suiltable for length compensation of heated columns and angular misalignment
 
  • Cone size:
    29/32 NS
  • Socket size:
    29/32 NS
  • Seq:
    04
  • Total length:
    120 mm