Order Entry
Belgium
Orders LinkContactUsLinkComponent
Dressing jars, with lid

Dressing jars, with lid

60 mm, 20 mm
Catalog # BOCH8760
Supplier:   BOCHEM
Dressing jars, with lid
Catalog # BOCH8760
Supplier:  BOCHEM
Dressing jars, with lid
Dressing jars, with lid

60 mm, 20 mm

Price
12,70 €
Log in for availability
1 * 1 ST
Product Details & Documents

Dressing jar either with flat or high shape, made of aluminium.

  • Very good temperature resistance
  • Good chemical resistance
  • Lightweight
Specifications
  • Ext.Ø:
    60 mm
  • Height:
    20 mm
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.