Order Entry
Belgium
Orders LinkContactUsLinkComponent
Ladle scoops

Ladle scoops

1400 ml, 350 mm, 180 mm
Catalog # 231-1106
Supplier:   BOCHEM
Ladle scoops
Catalog # 231-1106
Supplier:  BOCHEM
Ladle scoops
Ladle scoops

1400 ml, 350 mm, 180 mm

Price
135,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel 18/10, anti-magnetic and electropolished.

  • Complete with flat handle
  • Corrosion resistant
  • Excellent thermal resistance
Specifications
  • Ø:
    180 mm
  • Capacity:
    1400 ml
  • Length:
    350 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.