Order Entry
Belgium
Orders LinkContactUsLinkComponent
Ladle scoops

Ladle scoops

620 ml, 345 mm, 140 mm
Catalog # 231-1104
Supplier:   BOCHEM
Ladle scoops
Catalog # 231-1104
Supplier:  BOCHEM
Ladle scoops
Ladle scoops

620 ml, 345 mm, 140 mm

Price
49,30 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel 18/10, anti-magnetic and electropolished.

  • Complete with flat handle
  • Corrosion resistant
  • Excellent thermal resistance
Specifications
  • Ø:
    140 mm
  • Capacity:
    620 ml
  • Length:
    345 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.