Order Entry
Belgium
Orders LinkContactUsLinkComponent
Ladle scoops

Ladle scoops

220 ml, 315 mm, 100 mm
Catalog # 231-1103
Supplier:   BOCHEM
Ladle scoops
Catalog # 231-1103
Supplier:  BOCHEM
Ladle scoops
Ladle scoops

220 ml, 315 mm, 100 mm

Price
31,20 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel 18/10, anti-magnetic and electropolished.

  • Complete with flat handle
  • Corrosion resistant
  • Excellent thermal resistance
Specifications
  • Ø:
    100 mm
  • Capacity:
    220 ml
  • Length:
    315 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.