Order Entry
Belgium
Orders LinkContactUsLinkComponent
Powder funnels

Powder funnels

240 mm, 220 mm, 30 mm
Catalog # 221-1620
Supplier:   BOCHEM
Powder funnels
Catalog # 221-1620
Supplier:  BOCHEM
Powder funnels
Powder funnels

240 mm, 220 mm, 30 mm

Price
228,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel 18/10.

  • Smooth internal walls
  • Robust and strong
  • Good chemical resistance, extremely high thermal resistance

With short, wide stem.

Specifications
  • Funnel O-Ø:
    240 mm
  • Length:
    220 mm
  • Stem O-Ø:
    30 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.