Order Entry
Belgium
Orders LinkContactUsLinkComponent
Dressing jars, with lid

Dressing jars, with lid

100 ml, 60 mm, 60 mm
Catalog # BOCH8581
Supplier:   BOCHEM
Dressing jars, with lid
Catalog # BOCH8581
Supplier:  BOCHEM
Dressing jars, with lid
Dressing jars, with lid

100 ml, 60 mm, 60 mm

Price
34,90 €
Log in for availability
1 * 1 ST
Product Details & Documents

Jar made of stainless steel, with knob lid.

  • Very good temperature resistance
  • Good chemical resistance
  • Easy to clean
Specifications
  • Ext.Ø:
    60 mm
  • Capacity:
    100 ml
  • Height:
    60 mm