Instrument trays
440×340×20 mm
Catalog # BOCH8486
Supplier: BOCHEM
Jump to:
Product Details & Documents
Made of 18/10 steel.
- Very good chemical resistance
- Very good temperature resistance
- Sturdy
Documents
Certificates
Search by Lot NumberSpecifications
- W×D×H:440×340×20 mm