Orders LinkContactUsLinkComponent
Stand rods, with M10 thread
Stand rods, with M10 thread
12 mm, 750 mm
Catalog # 241-3159
Supplier:   BOCHEM
Stand rods, with M10 thread
Catalog # 241-3159
Supplier:  BOCHEM
Stand rods, with M10 thread
Stand rods, with M10 thread
12 mm, 750 mm
Price
15,70 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Aluminium, bare.
  • Lightweight, tough
  • With M10 thread
Spécifications
  • Ø:
    12 mm
  • Length:
    750 mm