Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Spoon spatulas

230 mm, 25×12 mm
Catalog # 231-1053
Supplier:   BOCHEM
Spoon spatulas
Catalog # 231-1053
Supplier:  BOCHEM
Spoon spatulas
Spoon spatulas

230 mm, 25×12 mm

Price
3,20 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel. Universal spoon spatula designed for a wide range of uses.

  • One end is shaped like a spoon and the other end like a spatula
  • With rounded handle
  • High thermal resistance, autoclavable
  • Excellent resistance to chemicals
  • Quick and easy to clean

 

Specifications
  • Length:
    230 mm
  • Spoon size:
    25×12 mm