Forceps with guide pin
Bent, sharp, 130 mm
Catalog # 232-0019
Supplier: BOCHEM
Jump to:
Product Details & Documents
Stainless steel, with PTFE coating.
- Bent, with guide pin
- Smooth, coated surface to prevent contamination and facilitate cleaning
Specifications
- Type:Bent, sharp
- Length:130 mm