Orders LinkContactUsLinkComponent
Forceps with guide pin
Forceps with guide pin
Sharp, straight, 145 mm
Catalog # 232-0017
Supplier:   BOCHEM
Forceps with guide pin
Catalog # 232-0017
Supplier:  BOCHEM
Forceps with guide pin
Forceps with guide pin
Sharp, straight, 145 mm
Price
36,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel, PTFE coating.
  • Anti-magnetic
  • Very good chemical and thermal resistance
  • Smooth surface prevents contamination and facilitates cleaning
Specifications
  • Type:
    Sharp, straight
  • Length:
    145 mm