Forceps with guide pin
Sharp, straight, 130 mm
Catalog # 232-2137
Supplier: BOCHEM
Jump to:
Product Details & Documents
Stainless steel, PTFE coating.
- Anti-magnetic
- Very good chemical and thermal resistance
- Smooth surface prevents contamination and facilitates cleaning
Specifications
- Type:Sharp, straight
- Length:130 mm