Orders LinkContactUsLinkComponent
Forceps, precision
Curved, 105 mm
  232-0008
 :   BOCHEM
Forceps, precision
  232-0008
 :  BOCHEM
Forceps, precision
Forceps, precision
Curved, 105 mm
10,00 €
1 * 1 ST
Stainless steel ¹⁸/₁₀, anti-magnetic. Extremely precise forceps designed for a wide variety of uses in the laboratory, production and more.
  • Extremely sharp, flat
  • Non serrated points
  • Type:
    Curved
  • Length:
    105 mm