Forceps, precision
Precision forceps, smooth tips, stainless steel, Straight, extra sharp, 120 mm
Catalog # BOCH1600
Supplier: BOCHEM
Jump to:
Product Details & Documents
These non serrated precision forceps are available in pure stainless steel or with PTFE coating.
Specifications
- Type:Straight, extra sharp
- Description:Precision forceps, smooth tips, stainless steel
- Length:120 mm