Weighing scoops
90 mm, 32 mm
Catalog # 611-9711
Supplier: BOCHEM
Aller à :
Product Details & Documents
Stainless steel 18/10, anti-magnetic.
Spécifications
- Width:32 mm
- Length:90 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



