Orders LinkContactUsLinkComponent
Forceps, sharp
Forceps, sharp
Sharp/bent, 145 mm
Catalog # 232-0059
Supplier:   BOCHEM
Forceps, sharp
Catalog # 232-0059
Supplier:  BOCHEM
Forceps, sharp
Forceps, sharp
Sharp/bent, 145 mm
Price
6,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel ¹⁸/₁₀, antimagnetic, bent.
Specifications
  • Type:
    Sharp/bent
  • Length:
    145 mm