Orders LinkContactUsLinkComponent
Forceps
Forceps
Straight, sharp, 105 mm
Catalog # BOCH1010
Supplier:   BOCHEM
Forceps
Catalog # BOCH1010
Supplier:  BOCHEM
Forceps
Forceps
Straight, sharp, 105 mm
Price
4,90 €
Log in for availability
1 * 1 ST
Product Details & Documents
Precision forceps made from stainless steel 18/10.
Documents
Specifications
  • Type:
    Straight, sharp
  • Length:
    105 mm
  • Seq:
    01