Orders LinkContactUsLinkComponent
Torso
Torso
Torso (Electronic Light Contr.)
Catalog # BINH1005618
Marketsource
Supplier:   3B Scientific
Torso
Catalog # BINH1005618
Supplier:  3B Scientific
Marketsource
Torso
Torso
Torso (Electronic Light Contr.)
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
This easy to transport torso allows the practice of adult CPR techniques. Optionally child CPR can also be practiced by changing the supplied springs inside the torso. Individual student face masks with a one-way non-rebreathing valve and disposable airways provide maximum protection against cross-contamination.
  • With light controller
Specifications
  • Description:
    Torso (Electronic Light Contr.)