Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Visual immunoassays
Visual immunoassays
Salmonella, VIA kit, 96 well, 96 Tests
Catalog # 709-0253
Visual immunoassays
Catalog # 709-0253
Supplier:  
CAS Number:  
Visual immunoassays
Visual immunoassays
Salmonella, VIA kit, 96 well, 96 Tests
CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Singlepath® Salmonella Test rapide pour la détermination de Salmonella dans les aliments 1 * 25 Tests Catalog #1.04140.0001
Product 1.04140.0001 is similar to 709-0253 but manufacturer and specifications may vary.
Product Details & Documents
Microtitre-format screening tests with high-affinity, specific antibodies for food and environment samples. Fast, specific bacteria and toxin testing with advanced Visual Immunoassay for food, water, environment samples, raw materials, cosmetics and pharmaceuticals. Ideal for rapid detection of pathogenic bacteria, 46 or 94 tests per plate.
  • The simplicity of the VIA™ method frees up time for other work and projects
  • Rapid results allow products to be released earlier, thereby saving on warehousing costs
  • The kit can be used manually, semi automatically or fully automatically for large numbers of samples
  • Fast results
  • High sensitivity and specificity
Specifications
Specifications
  • Cdt:
    96 Tests
  • Désignation:
    Salmonella, VIA kit, 96 well
Specifications