Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Polysorbate 80

Glass bottle, 250 mL
Catalog # ACRO278632500
Polysorbate 80
Catalog # ACRO278632500
CAS Number:  9005-65-6
Polysorbate 80
Polysorbate 80

Glass bottle, 250 mL

CAS Number:  9005-65-6
Price
41,00 €
Log in for availability
1 * 250 mL
Product Details & Documents
Synonyms:  Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate
  • CAS Number:9005-65-6
  • MDL Number:MFCD00082107
  • Merck Index:13,07664
  • Boiling Pt:? 100 °C (1013 hPa)
  • Melting Pt:-21 °C
  • Density:1.08 g/cm³
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • EINECS:500-019-9
Specifications

Specification Test Results

  • Appearance:
    Clear yellow to amber Viscous liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4700 to 1.4760 (20°C, 589 nm)
  • Water:
    ≤3 %
  • Acid value:
    ≤2 mg KOH/g
  • Hydroxyl value:
    65 to 80 mg KOH/g
  • Saponification value:
    45 to 55 mg KOH/g

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.