Order Entry
Belgium
Orders LinkContactUsLinkComponent
Methyl red, pure indicator

Methyl red, pure indicator

Glass bottle for solids, 25 g
Catalog # ACRO264440250
Methyl red, pure indicator
Catalog # ACRO264440250
CAS Number:  493-52-7
Methyl red, pure indicator
Methyl red, pure indicator

Glass bottle for solids, 25 g

CAS Number:  493-52-7
Price
42,20 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
  • CAS Number:493-52-7
  • Formula:C₁₅H₁₅N₃O₂
  • MDL Number:MFCD00002425
  • Molecular Weight:269.3 g/mol
  • Merck Index:12,06199
  • Melting Pt:178…182 °C
  • Density:1,31 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:207-776-1

Documents

Specifications

Specification Test Results

  • Appearance:
    Dark red to brown to purple to violet Powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    178 to 182 °C
  • Solubility:
    (0.1 % in ethanol) Clear
  • Specific extinction E (1%/1cm):
    ≥650 (430 nm, in buffer)
  • Visual transition interval:
    from pH 4.2 (pink or red) to pH 6.2 (yellow)

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.