Orders LinkContactUsLinkComponent
Cell scrapers
Cell scrapers
Small cell scraper, stainless steel, 15,88×6,35 mm blade, handle length 197 mm
Catalog # BELC7731-22000
Marketsource
Supplier:   BELLCO GLASS
Cell scrapers
Catalog # BELC7731-22000
Supplier:  BELLCO GLASS
Marketsource
Cell scrapers
Cell scrapers
Small cell scraper, stainless steel, 15,88×6,35 mm blade, handle length 197 mm
Price
140,01 €
Log in for availability
1 * 2 ST
Product Details & Documents
Stainless steel blade holder mounted on plastic covered stainless steel handle.
Specifications
  • Description:
    Small cell scraper, stainless steel, 15,88×6,35 mm blade, handle length 197 mm