Buffer solution pH 7, Phosphate buffer
Potassium dihydrogen phosphate and di-Sodium hydrogen phosphate in aqueous solution, 7,00, ±0,02, Phosphate, Plastic bottle, 1 L
Supplier: Thermo Fisher Scientific
Buffer solution pH 7, Phosphate buffer
Catalog # ACRO258590010
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
Documents
Specifications
Specifications
- Pack type:Plastic bottle
- pH buffer type:Phosphate
- Pk:1 L
- Description:Potassium dihydrogen phosphate and di-Sodium hydrogen phosphate in aqueous solution
- pH accuracy:±0,02
- pH value:7,00
- Cat. No.:ACRO258590010
- Concentration:3.54 g potassium dihydrogen phosphate and 14.7 g i-sodium hydrogen phosphate per litre
- Appearance:Clear colourless solution
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



