Buffer solution pH 4, citrate buffer
4,00, ±0,02, Citrate, Colourless, Plastic bottle, 1 L
Supplier: Thermo Fisher Scientific
Buffer solution pH 4, citrate buffer
Catalog # ACRO258580010
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
Documents
ACRO25858
SDS
Specifications
Specifications
- Pack type:Plastic bottle
- pH buffer type:Citrate
- Pk:1 L
- Colour:Colourless
- pH accuracy:±0,02
- pH value:4,00
- Cat. No.:ACRO258580010
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



