Order Entry
Belgium
Orders LinkContactUsLinkComponent
Polysorbate 20

Polysorbate 20

Glass bottle, 5 mL
Catalog # ACRO233360050
Polysorbate 20
Catalog # ACRO233360050
CAS Number:  9005-64-5
Polysorbate 20
Polysorbate 20

Glass bottle, 5 mL

CAS Number:  9005-64-5
Price
22,60 €
Log in for availability
1 * 5 mL
Product Details & Documents
Synonyms:  Polyoxyethylene-20-Sorbitan Monolaurate, Sorbitan monolaurate, ethoxylated
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Boiling Pt:>100 °C (760 mmHg)
  • Density:1.1 g/cm³
  • Flash Pt:>150 °C
  • Storage Temperature:Ambient
  • EINECS:500-018-3

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear to slightly hazy yellow to brown-yellow Viscous liquid (May develop some turbidity or precipitate)
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4680 to 1.4720 (20°C, 589 nm)
  • Water:
    ≤3 %
  • Dioxane:
    ≤10 ppm
  • Acid value:
    ≤2 mg KOH/g
  • Hydroxyl value:
    96 to 108 mg KOH/g
  • Saponification value:
    40 to 50 mg KOH/g

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL

Specifications