D-(+)-Biotin 98%
Glass bottle for solids, 500 mg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
- CAS Number:58-85-5
- Formula:C₁₀H₁₆N₂O₃S
- MDL Number:MFCD00005541
- Molecular Weight:244.31 g/mol
- Merck Index:13,01231
- Melting Pt:231…233 °C (dec.)
- Density:1,41 g/cm³ (20 °C)
- Storage Temperature:Refrigerator
- EINECS:200-399-3
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Loss on drying:≤1 % (105°C)
- Sulfated ash:≤0.1 %
- Appearance of solution:(2% soln. in 0.1 N NaOH) clear, colorless
- Specific optical rotation:+89° to +93° (20°C, 589 nm) (c=2, 0.1 N NaOH) (on dried substance)
- Titration with NaOH:≥97.5 % (On dried substance)
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg



