Creatinine 98%
Glass bottle, 50 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 2-Amino-1-methyl-1,5-dihydro-4H-imidazol-4-one
- CAS Number:60-27-5
- Formula:C₄H₇N₃O
- MDL Number:MFCD00059730
- Molecular Weight:113.12 g/mol
- Melting Pt:255 °C (dec.)
- Flash Pt:290 °C
- Storage Temperature:Ambient
- EINECS:200-466-7
Specifications
Specification Test Results
- Appearance:White to yellowish-white Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤1 % (105°C)
- Sulfated ash:≤0.2 %
- Solubility:(5 % in water) Clear colorless to slightly yellow
- Titration with HClO4:97.5 to 102.5 % (on dried substance)
- Heavy metals (as Pb):≤20 ppm
Specifications
- Pack type:Glass bottle
- Pk:50 g



