Order Entry
Belgium
Orders LinkContactUsLinkComponent
Creatinine 98%

Creatinine 98%

Glass bottle, 25 g
Catalog # ACRO228940250
Creatinine 98%
Catalog # ACRO228940250
CAS Number:  60-27-5
Creatinine 98%
Creatinine 98%

Glass bottle, 25 g

CAS Number:  60-27-5
Price
68,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  2-Amino-1-methyl-1,5-dihydro-4H-imidazol-4-one
  • CAS Number:60-27-5
  • Formula:C₄H₇N₃O
  • MDL Number:MFCD00059730
  • Molecular Weight:113.12 g/mol
  • Melting Pt:255 °C (dec.)
  • Flash Pt:290 °C
  • Storage Temperature:Ambient
  • EINECS:200-466-7

Documents

Specifications

Specification Test Results

  • Appearance:
    White to yellowish-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤1 % (105°C)
  • Sulfated ash:
    ≤0.2 %
  • Solubility:
    (5 % in water) Clear colorless to slightly yellow
  • Titration with HClO4:
    97.5 to 102.5 % (on dried substance)
  • Heavy metals (as Pb):
    ≤20 ppm

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.