Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Digoxin 95%

Glass bottle, 1 g
N° de catalogue ACRO228120010
Toxic
Fournisseur:   Thermo Fisher Scientific
Digoxin 95%
N° de catalogue ACRO228120010
Numéro CAS:  20830-75-5
Digoxin 95%
Digoxin 95%

Glass bottle, 1 g

Numéro CAS:  20830-75-5
Prix
115,00 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:20830-75-5
  • Formula:C₄₁H₆₄O₁₄
  • MDL Number:MFCD00003674
  • Molecular Weight:780.95 g/mol
  • Melting Pt:248…250 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:244-068-1
  • UN:3462
  • ADR:6.1,II
Specifications

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤1 % (500 mg) (vacuum)
  • Sulfated ash:
    ≤0.1 %
  • HPLC:
    ≥94 % (on dried substance)
  • Specific optical rotation:
    +10° to +13° (20°C, 589 nm) (c=2, anhydr. pyridine) (on dried substance)

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g

Specifications