Sample loops for Valco valves
Sample loop, for Valco W and UW type valves, Stainless steel, 250 µl
N° de catalogue JOURJR-SLV-76-250
Fournisseur: VICI JOUR RESEARCH
Aller à :
Product Details & Documents
These VICI Jour sample loops are made of high quality stainless steel 316.
- Fits all Valco W and UW type valves
- High pressure resistance, low permeability
- Tolerance of ±0,05 mm and pressure rating of <350 bar
They are cut using an electrolytic cutting process, cleaned with ultrasound and high pressure steam to remove contaminants, dried, and capped with softener-free caps to avoid recontamination of the tubing.
Livraison: Supplied with stainless hex-head nuts and stainless ferrules for reliable connections.
Spécifications
- Capacity:250 µl
- Description:Sample loop, for Valco W and UW type valves
- Material:Stainless steel
- Seq:06
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



