Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sample loops for Valco valves

Sample loops for Valco valves

Sample loop, for Valco W and UW type valves, Stainless steel, 1000 µl
Catalog # JOURJR-SLV-76-1000
Supplier:   VICI JOUR RESEARCH
Sample loops for Valco valves
Catalog # JOURJR-SLV-76-1000
Supplier:  VICI JOUR RESEARCH
Sample loops for Valco valves

Sample loop, for Valco W and UW type valves, Stainless steel, 1000 µl

Price
107,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These VICI Jour sample loops are made of high quality stainless steel 316.

  • Fits all Valco W and UW type valves
  • High pressure resistance, low permeability
  • Tolerance of ±0,05 mm and pressure rating of <350 bar

They are cut using an electrolytic cutting process, cleaned with ultrasound and high pressure steam to remove contaminants, dried, and capped with softener-free caps to avoid recontamination of the tubing.

Delivery information: Supplied with stainless hex-head nuts and stainless ferrules for reliable connections.

Specifications
  • Capacity:
    1000 µl
  • Description:
    Sample loop, for Valco W and UW type valves
  • Material:
    Stainless steel
  • Seq:
    08