Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sample loops for Valco valves

Sample loops for Valco valves

Sample loop, PEEK, 50 µl, max. 400 bar
Catalog # JOURJR-SLV-75-50
Supplier:   VICI JOUR RESEARCH
Sample loops for Valco valves
Catalog # JOURJR-SLV-75-50
Supplier:  VICI JOUR RESEARCH
Sample loops for Valco valves

Sample loop, PEEK, 50 µl, max. 400 bar

Price
75,30 €
Log in for availability
1 * 1 ST
Product Details & Documents

These sample loops are made from PEEK tubing and fittings. Mechanically strong, chemically inert polymer ideal for HPLC applications. Softener-free polyethylene caps are used to avoid contamination of the tubing.

  • Fits Valco W and UW type valves
  • Tolerance of ±0,025 mm

PEEK sample loops are particularly well suited for separations involving proteins, peptides, nucleic acids or other samples of biological origin as well as in IC systems where metal traces may interact with ionic samples or release transition metals that may deactivate ion exchange columns.

Delivery information: Supplied with PEEK hex-head nuts and PEEK ferrules.

Specifications
  • Capacity:
    50 µl
  • Description:
    Sample loop
  • Material:
    PEEK
  • Seq:
    04
  • Pressure:
    max. 400 bar
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.