Orders LinkContactUsLinkComponent
Dispersing elements for homogenisers
Dispersing elements for homogenisers
S 18 N - 19 G, T 18 basic, 15,9 m/s, 19,0/12,7 mm, 204 mm, 10 - 1500 ml
Catalog # 431-2603
Supplier:   IKA
Dispersing elements for homogenisers
Catalog # 431-2603
Supplier:  IKA
Dispersing elements for homogenisers
Dispersing elements for homogenisers
S 18 N - 19 G, T 18 basic, 15,9 m/s, 19,0/12,7 mm, 204 mm, 10 - 1500 ml
Price
1 350,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Type N: Materials in contact with medium are PTFE and stainless steel AISI 316L, suitable for solvents and abrasive substances, can be sterilised by all methods, max. temperature 180 °C.
Type NK: Materials in contact with medium are PTFE and stainless steel AISI 316L, suitable for solvents and abrasive substances, can be sterilised by chemical methods, max. temperature 120 °C.
Type KV: Materials in contact with medium are FFPM/SIC and stainless steel AISI 316L, suitable for vacuum down to 1 mbar, pressure up to 6 bar and for solvents, can be sterilised by chemical methods, max. temperature 220 °C.
SMK jet mixer head: The vertical flow and the high circumferential speed up to 10000 rpm ensure intensive mixing. The head is used for adding gases or liquids, for lump-free suspension of difficult to dissolve powders or for dissolving sedimented, already hardened substances.
Type KD: Materials in contact with the medium are PTFE, FFKM O-ring and stainless steel AISL 316L, suitable for solvents, can be sterilised by all methods, max. temperature 150 °C.
Caution: More dispersing elements are available on request. Please contact Avantor for details.
ST= sawtooth.
Specifications
  • Speed:
    15,9 m/s
  • Volume:
    10 - 1500 ml
  • Length:
    204 mm
  • Seq:
    07
  • Stator/rotor Ø:
    19,0/12,7 mm
  • Type:
    S 18 N - 19 G
  • For:
    T 18 basic
Accessories
Product Information
  • Catalog #
    For unit
    Availability
    Price
  • Bushing
    Bushing
    IKAA0150700
    S 18 N – 18 G
    Log in for availability
    1 * 1 ST - 145,00 €
Product Information
  • Catalog #
    For unit
    Availability
    Price
  • Driveshaft ring, PTFE
    Driveshaft ring, PTFE
    IKAAL004880
    S 18 N – 19 G
    Log in for availability
    1 * 1 ST - 17,50 €
Product Information
  • Catalog #
    For unit
    Availability
    Price
  • Rotor
    Rotor
    IKAA0158000
    S 18 N – 18 G
    Log in for availability
    1 * 1 ST - 192,00 €
Product Information
  • Catalog #
    For unit
    Availability
    Price
  • Stator
    Stator
    IKAA0161900
    S 18 N – 18 G
    Log in for availability
    1 * 1 ST - 609,00 €
Product Information
  • Catalog #
    For unit
    Availability
    Price
  • Silentstream flow breaker
    Silentstream flow breaker
    IKAA3754000
    S 25 N – 18 G; S 25 N – 25 G; S 25 N – 25 F; S 25 NK – 19 G; S 25 KV – 18 G; S 25 KV – 25 G; S 25 KV – 25 F; S 18 N – 19 G; S 25 N – 18 G - ST;S 25 N – 25 G - ST
    Log in for availability
    1 * 1 ST - 28,00 €
Product Information
  • Catalog #
    For unit
    Availability
    Price
  • Shaft
    Shaft
    IKAA0593700
    S 18 N – 18 G
    Log in for availability
    1 * 1 ST - 506,00 €
Product Information
  • Catalog #
    For unit
    Availability
    Price
  • Shaft sleeve
    Shaft sleeve
    IKAA0593600
    S 18 N – 18 G
    Log in for availability
    1 * 1 ST - 198,00 €
Product Information
  • Catalog #
    For unit
    Availability
    Price
  • Shaft tube
    Shaft tube
    IKAA0593500
    S 18 N – 18 G
    Log in for availability
    1 * 1 ST - 1 300,00 €
Product Information
  • Catalog #
    For unit
    Availability
    Price
  • Protective cap
    Protective cap
    IKAA2757500
    S 18 N – 18 G
    Log in for availability
    1 * 1 ST - 5,50 €
Product Information
  • Catalog #
    For unit
    Availability
    Price
  • Slit-ring bushing
    Slit-ring bushing
    IKAA1951200
    S 25 N – 18 G
    Log in for availability
    1 * 1 ST - 18,10 €