Orders LinkContactUsLinkComponent
Sieves
Sieves
Stainless steel sieve, 3,00 mm, Sieve MF 3.0
Catalog # 412-1135
Supplier:   IKA
Sieves
Catalog # 412-1135
Supplier:  IKA
Sieves
Sieves
Stainless steel sieve, 3,00 mm, Sieve MF 3.0
Price
370,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Interchangeable sieve for insertion into the grinding heads ensures maximum particle size filtering.
Specifications
  • Type:
    Sieve MF 3.0
  • Description:
    Stainless steel sieve, 3,00 mm
  • Seq:
    10