Orders LinkContactUsLinkComponent
Sieves
Sieves
Stainless steel sieve, 2,00 mm, Sieve MF 2.0
Catalog # 412-1138
Supplier:   IKA
Sieves
Catalog # 412-1138
Supplier:  IKA
Sieves
Sieves
Stainless steel sieve, 2,00 mm, Sieve MF 2.0
Price
384,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Interchangeable sieve for insertion into the grinding heads ensures maximum particle size filtering.
Specifications
  • Type:
    Sieve MF 2.0
  • Description:
    Stainless steel sieve, 2,00 mm
  • Seq:
    09